This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147137_m1
See other ZFP426 GE Assays ›
Gene Symbol:
Zfp426
Gene Name:
zinc finger protein 426
Gene Aliases:
2900057C04Rik, KRAB1, Zfo61, Zfp68-rs1, Znf426, mKIAA4237
Chromosome Location:
Chr.9: 20468548 - 20493922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147137_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp426
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110309.1 NP_001103779.1 4 - 5 346 86
NM_146221.4 NP_666333.1 2 - 3 484 86
GenBank mRNA AK085144.1 - 2 - 3 485 86
BC024805.1 - 4 - 5 342 5359610 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK085144.1, BC024805.1, NM_001110309.1, NM_146221.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp426

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0