This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147088_m1
See other DIS3L GE Assays ›
Gene Symbol:
Dis3l
Gene Name:
DIS3 mitotic control homolog (S. cerevisiae)-like
Gene Aliases:
AV340375
Chromosome Location:
Chr.9: 64306756 - 64341423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147088_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dis3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001295.2 NP_001001295.2 14 - 15 2664 132
NM_001177784.1 NP_001171255.1 14 - 15 2625 132
NM_172519.3 NP_766107.1 14 - 15 2568 132
GenBank mRNA AK029974.1 - 14 - 15 2569 132
AK032451.1 - 14 - 15 2572 132
AK077031.1 - 14 - 15 2628 132
AK150851.1 - 14 - 15 2610 132
AK151780.1 - 14 - 15 2610 132
AK153433.1 - 14 - 15 2610 132
AK173310.1 - 14 - 15 2598 132
BC056939.1 - 14 - 15 2597 5708685 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002492.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dis3l

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
mRNA catabolic process
rRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
RNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
hydrolase activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_001001295.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_001001295.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_001177784.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_001177784.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_172519.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 NM_172519.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900020909.mm.1 TC0900002492.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0