This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01144815_m1
See other STK11 GE Assays ›
Gene Symbol:
Stk11
Gene Name:
serine/threonine kinase 11
Gene Aliases:
AA408040, Lkb1, Par-4, R75140
Chromosome Location:
Chr.10: 80115693 - 80130685 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01144815_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stk11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB015801.1 - 8 - 9 1157 64
AF129870.1 - 8 - 9 1535 64
AF145287.1 - 8 - 9 1139 64
AF151711.1 - 8 - 9 1139 64
AK004499.1 - 6 - 7 818 64
AK172385.1 - 8 - 9 1922 64
AK172528.1 - 8 - 9 1919 64
BC052379.1 - 8 - 9 1200 5688695 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000839.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0