Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900018542.mm.1 |
NM_001289755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018542.mm.1 |
NM_001289756.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018542.mm.1 |
NM_023114.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018542.mm.1 |
TC0900002218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018549.mm.1 |
NM_001289756.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018549.mm.1 |
TC0900002218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018541.mm.1 |
NM_001289755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018541.mm.1 |
NM_001289756.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018541.mm.1 |
NM_001289833.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018541.mm.1 |
NM_023114.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018541.mm.1 |
TC0900002218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018543.mm.1 |
NM_001289755.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018543.mm.1 |
TC0900002218.mm.1 |
MTA TC |