This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01139069_g1
See other MVP GE Assays ›
Gene Symbol:
Mvp
Gene Name:
major vault protein
Gene Aliases:
2310009M24Rik, LRP, VAULT1
Chromosome Location:
Chr.7: 126986860 - 127014594 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01139069_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mvp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080638.3 NP_542369.2 9 - 10 1535 63
GenBank mRNA AF210456.1 - 9 - 10 1537 63
AK088046.1 - 9 - 10 1520 63
BC006709.1 - 9 - 10 1500 3584439 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004660.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mvp

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
negative regulation of signaling
negative regulation of protein autophosphorylation
ERBB signaling pathway
negative regulation of protein tyrosine kinase activity
protein activation cascade
protein kinase binding
protein phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700036111.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036111.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036111.mm.1 TC0700004660.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700036110.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036110.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036110.mm.1 TC0700004660.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0