This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01139044_m1
See other ANKHD1 GE Assays ›
Gene Symbol:
Ankhd1
Gene Name:
ankyrin repeat and KH domain containing 1
Gene Aliases:
1110004O12Rik, 4933432B13Rik, 9130019P20Rik, A530027J04Rik, A630021B20Rik, AA571404, Mask, mFLJ00246, mKIAA1085
Chromosome Location:
Chr.18: 36560603 - 36658908 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01139044_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ankhd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175375.3 NP_780584.2 9 - 10 1820 72
GenBank mRNA AK029322.1 - 2 - 3 235 72
AK131166.1 - 9 - 10 1683 72
CF742822.1 - 4 - 5 692 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000332.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ankhd1

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002510.mm.1 NM_175375.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002510.mm.1 NM_175375.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800002510.mm.1 TC1800000332.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800002511.mm.1 NM_175375.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002511.mm.1 NM_175375.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800002511.mm.1 TC1800000332.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header