This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138651_m1
See other PMPCB GE Assays ›
Gene Symbol:
Pmpcb
Gene Name:
peptidase (mitochondrial processing) beta
Gene Aliases:
3110004O18Rik, MPP11, MPPB, MPPP52
Chromosome Location:
Chr.5: 21737160 - 21757152 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138651_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pmpcb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028431.2 NP_082707.1 4 - 5 479 73
GenBank mRNA AK013995.1 - 4 - 5 480 73
AK152673.1 - 4 - 5 480 73
AK169417.1 - 4 - 5 436 73
BC038321.1 - 3 - 4 350 73
BC051216.1 - 3 - 4 286 73
BC060662.1 - 4 - 5 472 73
BC065095.1 - 4 - 5 472 73
BC087943.1 - 4 - 5 491 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000176.mm.1

Set Membership:

Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
catalytic activity
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001277.mm.1 NM_028431.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001277.mm.1 NM_028431.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500001277.mm.1 TC0500000176.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0