This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138604_g1
See other EGFL8 GE Assays ›
Gene Symbol:
Egfl8
Gene Name:
EGF-like domain 8
Gene Aliases:
Ng3
Chromosome Location:
Chr.17: 34613200 - 34616572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138604_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egfl8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152922.3 NP_690886.3 8 - 9 924 57
GenBank mRNA AK148723.1 - 9 - 10 835 57
AY635583.1 - 8 - 9 925 57
BC055829.1 - 4 - 5 1246 57
BC139484.1 - 8 - 9 893 8862506 57
BC141312.1 - 8 - 9 893 9055924 57
BC145632.1 - 8 - 9 893 9054200 57

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Egfl8

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 BC055829.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 NM_152922.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AK148723.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AY635583.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AK086281.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200457 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142370 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 BC055829.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 NM_152922.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AK148723.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AY635583.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AK086281.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154827 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154847 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154843 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154831 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154836 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154841 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154829 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154826 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154825 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154833 - -
Whole Mouse Genome G4122A A_51_P225112 BC141312.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P225112 NM_152922.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P225112 AK148723.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124539 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351463 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351455 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351454 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351429 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154834 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154782 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154796 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154840 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154826 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154839 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154845 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154835 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154843 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154831 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154826 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154833 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154825 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154837 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154847 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154838 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154822 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154836 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154840 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154834 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154832 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154827 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154829 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10444430 - -
Affymetrix Mouse Gene 1.0 ST Array 10450214 - -
Affymetrix Mouse Gene 1.0 ST Array 10450223 - -
Affymetrix Mouse Gene 1.0 ST Array 10450222 - -
Affymetrix Mouse Gene 1.0 ST Array 10450221 - -
Affymetrix Mouse Gene 1.0 ST Array 10450217 - -
Affymetrix Mouse Gene 1.0 ST Array 10450215 - -
Affymetrix Mouse Gene 1.0 ST Array 10450219 - -
Affymetrix Mouse Gene 1.0 ST Array 10450213 - -
Affymetrix Mouse Gene 1.0 ST Array 10450220 - -
Affymetrix Mouse Gene 1.0 ST Array 10450218 - -
Affymetrix Mouse Gene 1.0 ST Array 10450216 - -
Affymetrix HT MG-430 PM Array Plate 1447953_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10444420 - -
Affymetrix Mouse Gene 1.1 ST Array 10450212 - -
Mouse Genome 430 2.0 Array 1447953_at - -
Mouse Genome 430 2.0 Array 1447953_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 81701_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 81701_at - -
Murine Genome U74v2 Set 101345_at - -
Murine Genome U74v2 Set 101345_PM_at - -
Murine Genome U74v2 Set 167187_at - -
Murine Genome U74v2 Set 167187_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1447953_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1447953_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1170647 - -
Illumina mouse-6 v1.0 expression beadchip 360102 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000360102 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001170647 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2638839 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593189 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2638839 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2593189 - -
mouse-6 v1.1 (Illumina) scl50018.6.1_12-S - -
mouse-6 v1.1 (Illumina) scl0001741.1_77-S - -
Illumina Sentrix mouse-6.v1 bead array 360102 - -
Illumina Sentrix mouse-6.v1 bead array 1170647 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl50018.6.1_12-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001741.1_77-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360102 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170647 - -
MouseRef-8 v2 Expression BeadChip ILMN_2593188 - -
MouseRef-8 v2 Expression BeadChip ILMN_2593187 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593188 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593187 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0