This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138601_g1
See other EGFL8 GE Assays ›
Gene Symbol:
Egfl8
Gene Name:
EGF-like domain 8
Gene Aliases:
Ng3
Chromosome Location:
Chr.17: 34613200 - 34616572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138601_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egfl8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152922.3 NP_690886.3 5 - 6 514 74
GenBank mRNA AK148723.1 - 5 - 6 515 74
AY635583.1 - 5 - 6 515 74
BC055829.1 - 1 - 2 836 74
BC139484.1 - 5 - 6 483 8862506 74
BC141312.1 - 5 - 6 483 9055924 74
BC145632.1 - 5 - 6 483 9054200 74

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Egfl8

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0