This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01137327_m1
See other PPP1R9A GE Assays ›
Gene Symbol:
Ppp1r9a
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 9A
Gene Aliases:
2810430P21Rik, 4930518N04Rik, 5330407E15, A230094E16Rik, BB181831, NRB, mKIAA1222, neurabin-I
Chromosome Location:
Chr.6: 4902747 - 5165661 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01137327_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r9a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181595.3 NP_853626.1 7 - 8 2400 63
GenBank mRNA AB091828.1 - 7 - 8 2400 63
AK029737.1 - 7 - 8 1166 63
AK138313.1 - 7 - 8 2329 63
AY308065.1 - 6 - 7 2075 63
BC053748.1 - 7 - 8 2288 6331919 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000016.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r9a

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
positive regulation of neuron projection development
calcium-mediated signaling
neuron projection development
positive regulation of protein kinase activity
negative regulation of synaptic transmission
regulation of filopodium assembly
negative regulation of stress fiber assembly
positive regulation of dendritic spine development
cellular response to toxic substance
negative regulation of long-term synaptic potentiation
positive regulation of long term synaptic depression
protein binding
protein C-terminus binding
protein phosphatase 1 binding
protein kinase binding
protein domain specific binding
protein complex binding
protein homodimerization activity
actin filament binding
GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600000219.mm.1 NM_181595.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600000219.mm.1 NM_181595.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600000219.mm.1 TC0600000016.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0