Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700012174.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012174.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012174.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012182.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012182.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012182.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012170.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012170.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012170.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012172.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012172.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012172.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012177.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012177.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012177.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012175.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012175.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012175.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012180.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012180.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012180.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012176.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012176.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012176.mm.1 |
TC0700001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012178.mm.1 |
NM_019990.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012178.mm.1 |
NM_019990.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012178.mm.1 |
TC0700001430.mm.1 |
MTA TC |