This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01135202_g1
See other CD36 GE Assays ›
Gene Symbol:
Cd36
Gene Name:
CD36 antigen
Gene Aliases:
FAT, GPIV, Scarb3
Chromosome Location:
Chr.5: 17781690 - 17888959 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01135202_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159555.1 NP_001153027.1 5 - 6 890 161
NM_001159556.1 NP_001153028.1 5 - 6 750 161
NM_001159557.1 NP_001153029.1 6 - 7 801 161
NM_001159558.1 NP_001153030.1 6 - 7 917 161
NM_007643.4 NP_031669.3 5 - 6 893 161
GenBank mRNA AK052825.1 - 5 - 6 694 161
AK128908.1 - 6 - 7 694 161
AK150648.1 - 5 - 6 714 161
AK150865.1 - 1 - 2 88 161
AK151355.1 - 5 - 6 729 161
AK152904.1 - 3 - 4 400 161
AK172162.1 - 6 - 7 739 161
BC010262.1 - 5 - 6 702 3481681 161
L23108.1 - 5 - 6 711 161

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd36

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
long-chain fatty acid metabolic process
positive regulation of cell-matrix adhesion
pattern recognition receptor signaling pathway
negative regulation of systemic arterial blood pressure
fatty acid metabolic process
triglyceride metabolic process
transport
receptor-mediated endocytosis
phagocytosis, recognition
cell adhesion
signal transduction
cell surface receptor signaling pathway
nitric oxide mediated signal transduction
response to mechanical stimulus
negative regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
fatty acid transport
long-chain fatty acid transport
plasma membrane long-chain fatty acid transport
negative regulation of angiogenesis
fatty acid oxidation
lipid storage
cGMP-mediated signaling
positive regulation of blood coagulation
cholesterol transport
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
plasma lipoprotein particle clearance
low-density lipoprotein particle clearance
lipoprotein transport
negative regulation of transcription factor import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
apoptotic cell clearance
positive regulation of MAPK cascade
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
negative regulation of growth of symbiont in host
long-chain fatty acid import
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
low-density lipoprotein particle mediated signaling
positive regulation of phagocytosis, engulfment
positive regulation of macrophage cytokine production
cellular response to bacterial lipopeptide
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to hydroperoxide
regulation of removal of superoxide radicals
positive regulation of blood microparticle formation
positive regulation of reactive oxygen species metabolic process
low-density lipoprotein receptor activity
protein binding
high-density lipoprotein particle binding
lipid binding
low-density lipoprotein particle binding
transforming growth factor beta binding
thrombospondin receptor activity
oleic acid binding
lipoteichoic acid receptor activity
lipoprotein particle binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0