This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01135187_m1
See other HGF GE Assays ›
Gene Symbol:
Hgf
Gene Name:
hepatocyte growth factor
Gene Aliases:
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF
Chromosome Location:
Chr.5: 16553335 - 16622408 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01135187_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hgf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289458.1 NP_001276387.1 6 - 7 837 71
NM_001289459.1 NP_001276388.1 6 - 7 826 71
NM_001289461.1 NP_001276390.1 5 - 6 644 71
NM_010427.5 NP_034557.3 6 - 7 809 71
GenBank mRNA AK042121.1 - 6 - 7 755 71
BC119228.1 - 6 - 7 696 8733977 71
BC144764.1 - 6 - 7 708 71
D10212.1 - 5 - 6 632 71
D10213.1 - 5 - 6 617 71
S71816.1 - 5 - 6 632 71
X72307.1 - 5 - 6 693 71
X84046.1 - 5 - 6 672 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000125.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hgf

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
cell morphogenesis
liver development
proteolysis
cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of cell migration
positive regulation of myelination
cellular response to hepatocyte growth factor stimulus
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of angiogenesis
hepatocyte growth factor receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
myoblast proliferation
cell chemotaxis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
positive regulation of neuron projection regeneration
negative regulation of release of cytochrome c from mitochondria
negative regulation of hydrogen peroxide-mediated programmed cell death
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of DNA biosynthetic process
serine-type endopeptidase activity
growth factor activity
chemoattractant activity
identical protein binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0