This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01135044_g1
See other PMPCB GE Assays ›
Gene Symbol:
Pmpcb
Gene Name:
peptidase (mitochondrial processing) beta
Gene Aliases:
3110004O18Rik, MPP11, MPPB, MPPP52
Chromosome Location:
Chr.5: 21737160 - 21757152 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01135044_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pmpcb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028431.2 NP_082707.1 10 - 11 1256 104
GenBank mRNA AK013995.1 - 10 - 11 1257 104
AK152673.1 - 10 - 11 1258 105
AK169417.1 - 10 - 11 1213 104
BC038321.1 - 9 - 10 1127 104
BC051216.1 - 9 - 10 1063 104
BC060662.1 - 9 - 10 3768 104
BC065095.1 - 9 - 10 3768 104
BC087943.1 - 10 - 11 1268 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
catalytic activity
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0