This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01135019_m1
See other NUPL2 GE Assays ›
Gene Symbol:
Nupl2
Gene Name:
nucleoporin like 2
Gene Aliases:
AV116043, CG1, NLP-1, Nlp1
Chromosome Location:
Chr.5: 24164700 - 24184008 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01135019_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nupl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153092.4 NP_694732.3 4 - 5 635 140
GenBank mRNA AK077066.1 - 4 - 5 602 140
AK078478.1 - 4 - 5 636 140
BC033270.1 - 4 - 5 577 3981211 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000206.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nupl2

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
protein transport
mRNA transport
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001503.mm.1 NM_153092.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001503.mm.1 NM_153092.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500001503.mm.1 TC0500000206.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0