This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01134838_m1
See other ACTR3B GE Assays ›
Gene Symbol:
Actr3b
Gene Name:
ARP3 actin-related protein 3B
Gene Aliases:
9630005C02, ARP11, AW047569, Arp3b, Arp3beta
Chromosome Location:
Chr.5: 25759972 - 25850688 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01134838_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actr3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004365.1 NP_001004365.1 3 - 4 285 98
GenBank mRNA AK035798.1 - 2 - 3 197 98
BC053106.1 - 2 - 3 258 98
BC082279.1 - 3 - 4 285 6332588 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000238.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Actr3b

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
biological_process
Arp2/3 complex-mediated actin nucleation
nucleotide binding
molecular_function
actin binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001838.mm.1 NM_001004365.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500001838.mm.1 TC0500000238.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0