This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01133653_g1
See other TCAF1 GE Assays ›
Gene Symbol:
Tcaf1
Gene Name:
TRPM8 channel-associated factor 1
Gene Aliases:
2810407D09Rik, 3321401G04Rik, A230020K05Rik, AI848529, Fam115a
Chromosome Location:
Chr.6: 42667999 - 42710110 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01133653_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcaf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029930.2 NP_084206.1 4 - 5 1848 98
GenBank mRNA AK014379.1 - 4 - 5 1377 98
AK053509.1 - 4 - 5 1849 98
AK076242.1 - 4 - 5 1794 98
AK083917.1 - 4 - 5 1796 98
AK132148.1 - 4 - 5 1816 98
AK148593.1 - 4 - 5 1827 98
AK173013.1 - 4 - 5 1830 98
BC031718.1 - 4 - 5 1876 3156550 98
BC043304.1 - 2 - 3 108 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002178.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcaf1

Gene Ontology Categories:

Function(s) Process(es)

transport
negative regulation of cell migration
positive regulation of protein targeting to membrane
positive regulation of anion channel activity
ion channel binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017179.mm.1 NM_029930.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017179.mm.1 NM_029930.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017179.mm.1 TC0600002178.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017180.mm.1 NM_029930.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017180.mm.1 NM_029930.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017180.mm.1 TC0600002178.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0