This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01132397_g1
See other DXO GE Assays ›
Gene Symbol:
Dxo
Gene Name:
decapping exoribonuclease
Gene Aliases:
Dom3z, NG6
Chromosome Location:
Chr.17: 34837019 - 34839186 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01132397_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dxo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163770.1 NP_001157242.1 6 - 7 1246 66
NM_033613.2 NP_291091.2 6 - 7 1271 66
GenBank mRNA BC004713.1 - 6 - 7 1207 3499698 66
BG071924.2 - 1 - 2 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dxo

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
metabolic process
RNA destabilization
nuclear mRNA surveillance
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
mRNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
thiamine-pyrophosphatase activity
5'-3' exonuclease activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
RNA pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156009 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156014 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156010 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156012 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156011 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156013 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156008 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 BC004713.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 NM_001163770.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 BC004713.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 NM_001163770.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499805 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125912 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 BC004713.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 NM_001163770.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 NM_033613.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 BC004713.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 NM_001163770.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 BC004713.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 NM_001163770.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 BC004713.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 NM_001163770.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156021 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156020 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156025 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156027 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156016 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156023 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156008 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31156011 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125912 - -
Whole Mouse Genome G4122A A_51_P267388 BC004713.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P267388 NM_001163770.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P267388 NM_033613.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P267388 BC004713.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P267388 NM_001163770.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P267388 NM_033613.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112868 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138083 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125912 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174878 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174868 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174886 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174879 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156009 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156010 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156011 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156014 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156012 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156013 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31156008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156027 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31156011 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13174879 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156023 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13174877 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156018 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13174883 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156008 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156025 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156020 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156027 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31156009 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10444499 - -
Affymetrix Mouse Gene 1.0 ST Array 10444501 - -
Affymetrix Mouse Gene 1.0 ST Array 10444503 - -
Affymetrix Mouse Gene 1.0 ST Array 10444497 - -
Affymetrix Mouse Gene 1.0 ST Array 10444502 - -
Affymetrix Mouse Gene 1.0 ST Array 10444504 - -
Affymetrix Mouse Gene 1.0 ST Array 10444498 - -
Affymetrix HT MG-430 PM Array Plate 1423116_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10444496 - -
Mouse Genome 430 2.0 Array 1423116_PM_at - -
Mouse Genome 430 2.0 Array 1423116_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 112403_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 112403_at - -
Affymetrix Mouse Clariom S Array PSR1700006393.mm.1 NM_001163770.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006393.mm.1 NM_001163770.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006393.mm.1 NM_033613.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006393.mm.1 NM_033613.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700006393.mm.1 TC1700000646.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006394.mm.1 TC1700000646.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006390.mm.1 NM_001163770.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006390.mm.1 NM_001163770.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006390.mm.1 NM_033613.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006390.mm.1 NM_033613.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700006390.mm.1 TC1700000646.mm.1 MTA TC
Mouse Expression Set 430 1423116_PM_at - -
Mouse Expression Set 430 1423116_at - -
Murine Genome U74v2 Set 97428_PM_at - -
Murine Genome U74v2 Set 97428_at - -
Murine Genome U74v2 Set 171165_r_at - -
Murine Genome U74v2 Set 171165_PM_r_at - -
Mouse Genome 430A 2.0 Array 1423116_PM_at - -
Mouse Genome 430A 2.0 Array 1423116_at - -
Affymetrix HT Mouse Genome 430A Array 1423116_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1423116_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 447256 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2360068 - -
MouseWG-6 v2 Expression BeadChip 3870358 - -
Illumina mouse-6 v1.0 expression beadchip 2360068 - -
Illumina mouse-6 v1.0 expression beadchip 3870358 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003870358 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002360068 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694156 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653508 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2694156 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2653508 - -
mouse-6 v1.1 (Illumina) scl50807.7.1_59-S - -
mouse-6 v1.1 (Illumina) scl0001747.1_14-S - -
Illumina Sentrix mouse-6.v1 bead array 2360068 - -
Illumina Sentrix mouse-6.v1 bead array 3870358 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001747.1_14-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl50807.7.1_59-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870358 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360068 - -
MouseRef-8 v2 Expression BeadChip ILMN_1233838 - -
MouseRef-8 v2 Expression BeadChip ILMN_2694156 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233838 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694156 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001747.1_14-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50807.7.1_59-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0