This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01132063_m1
See other PHTF2 GE Assays ›
Gene Symbol:
Phtf2
Gene Name:
putative homeodomain transcription factor 2
Gene Aliases:
1110054G21Rik, 9530062N20Rik, AI447096, C77923
Chromosome Location:
Chr.5: 20758664 - 20882124 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01132063_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phtf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172992.3 NP_766580.3 15 - 16 2202 133
GenBank mRNA AK037021.1 - 15 - 16 2089 133
AK163376.1 - 15 - 16 2129 133
BC052841.1 - 15 - 16 2115 30050876 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002007.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phtf2

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500018215.mm.1 NM_172992.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500018215.mm.1 NM_172992.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500018215.mm.1 TC0500002007.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500018214.mm.1 NM_172992.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500018214.mm.1 NM_172992.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500018214.mm.1 TC0500002007.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0