This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01131379_m1
See other KLHL35 GE Assays ›
Gene Symbol:
Klhl35
Gene Name:
kelch-like 35
Gene Aliases:
2810406K13Rik
Chromosome Location:
Chr.7: 99464216 - 99474022 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01131379_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Klhl35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028145.1 NP_082421.1 2 - 3 956 89
GenBank mRNA AK013012.1 - 2 - 3 934 89
BE994913.1 - 1 - 2 361 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001387.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700011747.mm.1 NM_028145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011747.mm.1 NM_028145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011747.mm.1 TC0700001387.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700011748.mm.1 NM_028145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011748.mm.1 NM_028145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011748.mm.1 TC0700001387.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700011746.mm.1 NM_028145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011746.mm.1 NM_028145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011746.mm.1 TC0700001387.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700011750.mm.1 NM_028145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011750.mm.1 NM_028145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011750.mm.1 TC0700001387.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0