This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01131097_m1
See other USP15 GE Assays ›
Gene Symbol:
Usp15
Gene Name:
ubiquitin specific peptidase 15
Gene Aliases:
4921514G19Rik, AI327321, E430033I05Rik, Gcap18
Chromosome Location:
Chr.10: 123105006 - 123197019 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01131097_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Usp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301628.1 NP_001288557.1 10 - 11 1490 153
NM_027604.4 NP_081880.2 11 - 12 1577 153
GenBank mRNA AF468037.1 - 11 - 12 1481 153
AK014891.1 - 5 - 6 788 153
AK122301.1 - 10 - 11 1405 153
AK133852.1 - 10 - 11 1399 153
AK145749.1 - 11 - 12 1481 153
AK161469.1 - 11 - 12 1520 153
AK161957.1 - 10 - 11 1438 153
AK163315.1 - 10 - 11 1419 153
AK168755.1 - 10 - 11 1425 153
BC050042.1 - 11 - 12 1493 30022307 153

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Usp15

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ubiquitin-dependent protein catabolic process
transforming growth factor beta receptor signaling pathway
protein deubiquitination
BMP signaling pathway
monoubiquitinated protein deubiquitination
histone H2B conserved C-terminal lysine deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
pathway-restricted SMAD protein phosphorylation
regulation of proteasomal protein catabolic process
catalytic activity
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
transforming growth factor beta receptor binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
identical protein binding
SMAD binding
ubiquitinated histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header