This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01130957_m1
See other PSMA1 GE Assays ›
Gene Symbol:
Psma1
Gene Name:
proteasome (prosome, macropain) subunit, alpha type 1
Gene Aliases:
C2, HC2, Pros-30, alpha-type
Chromosome Location:
Chr.7: 114264550 - 114276116 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01130957_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psma1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011965.2 NP_036095.1 1 - 2 116 75
GenBank mRNA AF060088.1 - 1 - 2 40 75
AJ272019.1 - 1 - 2 69 75
AK088673.1 - 1 - 2 86 75
AK135326.1 - 1 - 2 54 75
AK137054.1 - 1 - 2 58 75
AK144784.1 - 1 - 2 71 75
AK161611.1 - 1 - 2 58 75
AK162277.1 - 1 - 2 63 75
AK167689.1 - 1 - 2 54 75
BC005762.1 - 1 - 2 53 2655483 75
BY108158.1 - 1 - 2 116 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF060088.1, AJ272019.1, AK088673.1, AK137054.1, AK162277.1, AK167689.1, BC005762.1, NM_011965.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psma1

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

immune system process
negative regulation of inflammatory response to antigenic stimulus
proteolysis
ubiquitin-dependent protein catabolic process
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
lipopolysaccharide binding
endopeptidase activity
threonine-type endopeptidase activity
protein binding
peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700034286.mm.1 NM_011965.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034286.mm.1 NM_011965.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700034286.mm.1 TC0700004135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034284.mm.1 NM_011965.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034284.mm.1 NM_011965.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700034284.mm.1 TC0700004135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034290.mm.1 NM_011965.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034290.mm.1 NM_011965.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700034290.mm.1 TC0700004135.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0