This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01130465_m1
See other APC GE Assays ›
Gene Symbol:
Apc
Gene Name:
adenomatosis polyposis coli
Gene Aliases:
AI047805, AU020952, AW124434, CC1, Min, mAPC
Chromosome Location:
Chr.18: 34220307 - 34322190 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01130465_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007462.3 NP_031488.2 3 - 4 519 137
GenBank mRNA AK042021.1 - 3 - 4 520 137
AK045053.1 - 3 - 4 490 137
AK080907.1 - 3 - 4 519 137
AK082724.1 - 3 - 4 265 137
BC141141.1 - 2 - 3 232 9055753 137
M88127.1 - 2 - 3 208 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000282.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apc

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
kidney development
hair follicle development
protein complex assembly
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle arrest
metaphase/anaphase transition of mitotic cell cycle
mitotic spindle assembly checkpoint
establishment or maintenance of cell polarity
pattern specification process
axonogenesis
negative regulation of cell proliferation
axis specification
anterior/posterior pattern specification
dorsal/ventral pattern formation
proximal/distal pattern formation
Wnt signaling pathway
cell migration
stem cell population maintenance
negative regulation of Wnt signaling pathway
regulation of cell migration
positive regulation of cell migration
regulation of epithelial cell differentiation
positive regulation of epithelial cell differentiation
positive regulation of microtubule polymerization
cytoplasmic microtubule organization
neuron projection development
positive regulation of pseudopodium assembly
regulation of microtubule-based process
T cell differentiation in thymus
somatic stem cell population maintenance
negative regulation of odontogenesis
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAPK cascade
skin development
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
regulation of cell differentiation
positive regulation of cell differentiation
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell adhesion
muscle cell cellular homeostasis
thymus development
negative regulation of epithelial cell proliferation
regulation of nitrogen compound metabolic process
chromosome organization
regulation of cell cycle
positive regulation of cell division
regulation of attachment of spindle microtubules to kinetochore
retina development in camera-type eye
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
negative regulation of canonical Wnt signaling pathway
regulation of glutamate metabolic process
protease binding
protein binding
beta-catenin binding
microtubule binding
protein kinase regulator activity
protein kinase binding
protein complex binding
gamma-catenin binding
microtubule plus-end binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002022.mm.1 NM_007462.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002022.mm.1 NM_007462.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800002022.mm.1 TC1800000282.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0