Assay Details
Gene Symbol: Ier5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034055.1 | - | 2432 | 111 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103440 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103419 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103437 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103447 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103454 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103441 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103418 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103435 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103439 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103436 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103452 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103438 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103442 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103453 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103450 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121856 | BC040084.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121856 | AK034055.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121856 | BC003464.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121856 | BC010234.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121856 | NM_030244.3 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113078 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121856 | BC040084.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121856 | AK034055.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121856 | BC003464.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121856 | BC010234.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121856 | NM_030244.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21103467 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113078 | - | - |
Whole Mouse Genome G4122A | A_52_P562641 | AK034055.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P362514 | BC040084.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P362514 | AK034055.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P362514 | BC003464.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P362514 | BC010234.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P362514 | NM_030244.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110506 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113078 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01604750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103439 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103453 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103442 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103419 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103438 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103440 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103435 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103441 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103454 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103447 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103452 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103437 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103436 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21103418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21103407 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01771726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01551693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01705201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01771725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02146065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01604749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01604759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01179980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02177406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01771752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01604761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21103374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01604742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02194575 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10481493 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481494 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481495 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455881_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419066_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10481491 | - | - |
Mouse Genome 430 2.0 Array | 1455881_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455881_at | - | - |
Mouse Genome 430 2.0 Array | 1419066_at | - | - |
Mouse Genome 430 2.0 Array | 1419066_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72500_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72500_PM_at | - | - |
Mouse Expression Set 430 | 1419066_at | - | - |
Mouse Expression Set 430 | 1419066_PM_at | - | - |
Mouse Expression Set 430 | 1455881_at | - | - |
Mouse Expression Set 430 | 1455881_PM_at | - | - |
Murine Genome U74v2 Set | 171397_PM_at | - | - |
Murine Genome U74v2 Set | 171397_at | - | - |
Mouse Genome 430A 2.0 Array | 1419066_at | - | - |
Mouse Genome 430A 2.0 Array | 1419066_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419066_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419066_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455881_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455881_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 388521 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2450154 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002450154 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770609 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2770609 | - | - |
mouse-6 v1.1 (Illumina) | scl19470.2_417-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2450154 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19470.2_417-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450154 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2961005 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2961005 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770609 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19470.2_417-S | - | - |
