This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805581_m1
See other IGF2BP2 GE Assays ›
Gene Symbol:
Igf2bp2
Gene Name:
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases:
C330012H03Rik, IMP-2, Imp2, Neilsen
Chromosome Location:
Chr.16: 22059009 - 22163299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805581_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf2bp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183029.2 NP_898850.2 6 - 7 990 76
GenBank mRNA AK044984.1 - 5 - 6 506 76
AK049196.1 - 6 - 7 687 76
AK077404.1 - 6 - 7 670 76
AK159654.1 - 6 - 7 723 76
AK170531.1 - 5 - 6 494 76
AY531659.1 - 6 - 7 654 76
BC023758.1 - 6 - 7 688 76
BC054552.1 - 6 - 7 710 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001375.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igf2bp2

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
transport
mRNA transport
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
poly(A) RNA binding
mRNA 5'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600011391.mm.1 NM_183029.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011391.mm.1 NM_183029.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600011391.mm.1 TC1600001375.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0