This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00805339_g1
See other FGB GE Assays ›
Gene Symbol:
Fgb
Gene Name:
fibrinogen beta chain
Gene Aliases:
2510049G14Rik
Chromosome Location:
Chr.3: 83042303 - 83049790 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00805339_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181849.2 NP_862897.1 6 - 7 961 74
GenBank mRNA AF413205.1 - 2 - 3 233 74
AK011118.1 - 6 - 7 980 74
AK011920.1 - 6 - 7 976 74
AK146960.1 - 6 - 7 981 74
AK168626.1 - 6 - 7 975 74
BC031715.1 - 6 - 7 961 4195310 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgb

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cell-matrix adhesion
signal transduction
blood coagulation
hemostasis
platelet activation
positive regulation of heterotypic cell-cell adhesion
induction of bacterial agglutination
cellular protein complex assembly
innate immune response
positive regulation of vasoconstriction
positive regulation of exocytosis
positive regulation of protein secretion
protein polymerization
response to calcium ion
positive regulation of ERK1 and ERK2 cascade
platelet aggregation
positive regulation of peptide hormone secretion
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of endothelial cell apoptotic process
receptor binding
structural molecule activity
protein binding, bridging
cell adhesion molecule binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0