Assay Details
Gene Symbol: Ifitm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053088.2 | NP_444318.1 | 1 - 2 | 220 | 73 | |
GenBank mRNA | AJ009781.1 | - | 1 - 2 | 220 | 73 | |
BC103790.1 | - | 1 - 2 | 220 | 30287396 | 73 | |
EU380257.1 | - | 1 - 2 | 192 | 73 | ||
FJ200451.1 | - | 1 - 2 | 272 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846742 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110423 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | AK003363.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | BC103790.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | EU380257.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | AK003363.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | BC103790.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585845 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25585856 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110423 | - | - |
Whole Mouse Genome G4122A | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | NM_053088.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | AJ009781.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | AK003363.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | BC103790.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P273812 | EU380257.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135142 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110423 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05936244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06354421 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05936233 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05936237 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06354423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585873 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05936256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585827 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585846 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06354421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06354424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06653983 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06653976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06654003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25585854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06653988 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10569013 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569012 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440216_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10569011 | - | - |
Mouse Genome 430 2.0 Array | 1440216_at | - | - |
Mouse Genome 430 2.0 Array | 1440216_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73835_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73835_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700037894.mm.1 | NM_053088.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037894.mm.1 | NM_053088.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037894.mm.1 | TC0700004512.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700037893.mm.1 | NM_053088.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037893.mm.1 | NM_053088.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037893.mm.1 | TC0700004512.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430B Array | 1440216_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440216_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 706827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2360161 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002360161 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245864 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245864 | - | - |
mouse-6 v1.1 (Illumina) | scl30507.4.1_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2360161 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30507.4.1_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360161 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1245864 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245864 | - | - |
