This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00804054_m1
See other STK32B GE Assays ›
Gene Symbol:
Stk32b
Gene Name:
serine/threonine kinase 32B
Gene Aliases:
2510009F08Rik, STKG6, Stk32, YANK2
Chromosome Location:
Chr.5: 37446825 - 37717153 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00804054_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stk32b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022416.2 NP_071861.1 10 - 11 1406 70
GenBank mRNA AJ250840.1 - 10 - 11 1484 70
AK082468.1 - 10 - 11 1407 70
BC052404.1 - 10 - 11 1316 5715729 70
BC056396.1 - 10 - 11 1392 6841272 70
BC058412.1 - 10 - 11 1316 5715729 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002274.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stk32b

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020970.mm.1 NM_022416.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020970.mm.1 NM_022416.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500020970.mm.1 TC0500002274.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0