This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803740_g1
See other PSMA1 GE Assays ›
Gene Symbol:
Psma1
Gene Name:
proteasome (prosome, macropain) subunit, alpha type 1
Gene Aliases:
C2, HC2, Pros-30, alpha-type
Chromosome Location:
Chr.7: 114264550 - 114276116 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803740_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psma1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011965.2 NP_036095.1 8 - 9 732 82
GenBank mRNA AF060088.1 - 8 - 9 656 82
AJ272019.1 - 8 - 9 685 82
AK088673.1 - 8 - 9 699 82
AK137054.1 - 8 - 9 674 82
AK144784.1 - 8 - 9 688 82
AK161611.1 - 8 - 9 673 82
AK162277.1 - 8 - 9 679 82
AK167689.1 - 8 - 9 670 82
BC005762.1 - 8 - 9 669 2655483 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psma1

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

immune system process
negative regulation of inflammatory response to antigenic stimulus
proteolysis
ubiquitin-dependent protein catabolic process
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
lipopolysaccharide binding
endopeptidase activity
threonine-type endopeptidase activity
protein binding
peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0