This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803696_m1
See other MAP2K6 GE Assays ›
Gene Symbol:
Map2k6
Gene Name:
mitogen-activated protein kinase kinase 6
Gene Aliases:
MEK6, MKK6, Prkmk6, SAPKK3
Chromosome Location:
Chr.11: 110399102 - 110516212 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803696_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Map2k6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011943.2 NP_036073.1 2 - 3 370 62
GenBank mRNA AK043482.1 - 2 - 3 371 62
AK077712.1 - 2 - 3 365 62
AK086722.1 - 371 62
AK138757.1 - 2 - 3 371 62
AK140289.1 - 3 - 4 355 62
AK141708.1 - 2 - 3 355 62
AK162671.1 - 2 - 3 324 62
BC075652.1 - 2 - 3 369 30541969 62
U39066.1 - 2 - 3 284 62
X97052.1 - 2 - 3 228 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the U39066.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Map2k6

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
positive regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
regulation of mitotic cell cycle
regulation of signal transduction
phosphorylation
stress-activated protein kinase signaling cascade
positive regulation of prostaglandin secretion
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of nitric-oxide synthase biosynthetic process
cardiac muscle contraction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100017315.mm.1 NM_011943.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100017315.mm.1 NM_011943.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100017315.mm.1 TC1100001841.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header