This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00803124_m1
See other OGDH GE Assays ›
Gene Symbol:
Ogdh
Gene Name:
oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide)
Gene Aliases:
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192
Chromosome Location:
Chr.11: 6291593 - 6359094 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00803124_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252282.1 NP_001239211.1 2 - 3 347 71
NM_001252283.1 NP_001239212.1 2 - 3 347 71
NM_001252287.1 NP_001239216.1 2 - 3 423 71
NM_001252288.1 NP_001239217.1 2 - 3 407 71
NM_010956.4 NP_035086.2 2 - 3 347 71
GenBank mRNA AK147289.1 - 2 - 3 348 71
AK150009.1 - 2 - 3 313 71
AK169286.1 - 2 - 3 339 71
AK195097.1 - 1 - 2 127 71
AK202141.1 - 128 71
AK202346.1 - 1 - 2 127 71
AK220536.1 - 2 - 3 309 71
BC013670.1 - 2 - 3 281 4188988 71
BC049104.1 - 2 - 3 360 6417392 71
BC057354.1 - 2 - 3 275 6827509 71
BC061232.1 - 2 - 3 303 71
BY768178.1 - 2 - 3 340 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC061232.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogdh

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
tricarboxylic acid cycle
2-oxoglutarate metabolic process
succinyl-CoA metabolic process
NADH metabolic process
metabolic process
oxidation-reduction process
oxoglutarate dehydrogenase (succinyl-transferring) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
thiamine pyrophosphate binding
heat shock protein binding
oxoglutarate dehydrogenase (NAD+) activity
metal ion binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0