This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00802443_m1
See other DDX4 GE Assays ›
Gene Symbol:
Ddx4
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases:
AV206478, Mvh, VASA
Chromosome Location:
Chr.13: 112598333 - 112652629 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00802443_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145885.1 NP_001139357.1 17 - 18 1520 94
NM_010029.2 NP_034159.1 16 - 17 1442 94
GenBank mRNA AK014844.1 - 16 - 17 1433 94
AK133366.1 - 17 - 18 1513 94
BC137601.1 - 16 - 17 1496 8860620 94
BC144760.1 - 16 - 17 1496 9053308 94
JQ923486.1 - 16 - 17 1470 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx4

Gene Ontology Categories:

Function(s) Process(es)

male meiosis I
multicellular organism development
spermatogenesis
RNA secondary structure unwinding
sperm motility
regulation of protein localization
nucleotide binding
nucleic acid binding
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0