This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00801421_m1
See other ACO1 GE Assays ›
Gene Symbol:
Aco1
Gene Name:
aconitase 1
Gene Aliases:
Aco-1, Irebp, Irp1
Chromosome Location:
Chr.4: 40143265 - 40199009 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00801421_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aco1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007386.2 NP_031412.2 3 - 4 570 67
GenBank mRNA AK027914.1 - 3 - 4 356 67
AK085951.1 - 3 - 4 319 67
AK129025.1 - 3 - 4 352 67
AK163985.1 - 3 - 4 369 67
AK216262.1 - 2 - 3 281 67
BC005454.1 - 3 - 4 330 3494686 67
X61147.1 - 3 - 4 337 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aco1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
citrate metabolic process
regulation of translation
cellular iron ion homeostasis
metabolic process
post-embryonic development
response to iron(II) ion
regulation of gene expression
intestinal absorption
RNA binding
aconitate hydratase activity
lyase activity
iron-responsive element binding
metal ion binding
mRNA 5'-UTR binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0