Assay Details
Gene Symbol: N4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC004022.1 | - | 1 - 2 | 897 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | BC004022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | BC089324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | AK172981.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | AK147522.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | BC004022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | BC089324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | AK172981.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | AK147522.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643347 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67395 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265407 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245773 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111394 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547256 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218891 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301575 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526598 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559459 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861892 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969975 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361663 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672736 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745271 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932833 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699913 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431237 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588275 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488440 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396908 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558870 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548529 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520828 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470228 | AK016804.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372555 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83589 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178889 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60379 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108481 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49201 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172291 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113183 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102303 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109780 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160410 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146924 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | BC004022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | BC089324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | AK172981.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | AK147522.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | BC004022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | BC089324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | AK172981.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | AK147522.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109780 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113183 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102303 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06583765 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146924 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160410 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172291 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01787177 | - | - |
Whole Mouse Genome G4122A | A_52_P420216 | BC004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | BC089324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | AK172981.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | AK147522.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | BC004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | BC089324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | AK172981.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | AK147522.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420216 | NM_030563.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310773 | BC004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P199788 | AK053991.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108992 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146924 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172291 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102303 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109780 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160410 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07207036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031825 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031823 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06757509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06840442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07324629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06792706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07115621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07207032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07309146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07149877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07207035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06841705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07087632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031591 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07207028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06840441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07407604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07309143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06757505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26031718 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10580458 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580459 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580460 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580465 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580462 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580464 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580466 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580463 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580461 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455606_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446956_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418154_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446957_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10580457 | - | - |
Mouse Genome 430 2.0 Array | 1455606_at | - | - |
Mouse Genome 430 2.0 Array | 1455606_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418154_at | - | - |
Mouse Genome 430 2.0 Array | 1418154_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80750_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80750_at | - | - |
Mouse Expression Set 430 | 1418154_PM_at | - | - |
Mouse Expression Set 430 | 1418154_at | - | - |
Mouse Expression Set 430 | 1455606_at | - | - |
Mouse Expression Set 430 | 1455606_PM_at | - | - |
Murine Genome U74v2 Set | 103930_PM_at | - | - |
Murine Genome U74v2 Set | 103930_at | - | - |
Mouse Genome 430A 2.0 Array | 1418154_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418154_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418154_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418154_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446957_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455606_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455606_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446956_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446957_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446956_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 663205 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106180072 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106180072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621901 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2621901 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1239821 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106180072 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106180072 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2570913 | - | - |
