Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800019367.mm.1 |
NM_001170691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019367.mm.1 |
NM_001170691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019367.mm.1 |
NM_178736.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019367.mm.1 |
NM_178736.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019367.mm.1 |
TC0800002550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019363.mm.1 |
NM_001170691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019363.mm.1 |
NM_001170691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019363.mm.1 |
NM_178736.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019363.mm.1 |
NM_178736.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019363.mm.1 |
TC0800002550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019366.mm.1 |
NM_001170691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019366.mm.1 |
NM_001170691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019366.mm.1 |
NM_178736.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019366.mm.1 |
NM_178736.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019366.mm.1 |
TC0800002550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019368.mm.1 |
NM_001170691.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019368.mm.1 |
NM_001170691.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019368.mm.1 |
NM_178736.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019368.mm.1 |
NM_178736.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019368.mm.1 |
TC0800002550.mm.1 |
MTA TC |