This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00724026_m1
See other GEN1 GE Assays ›
Gene Symbol:
Gen1
Gene Name:
Gen homolog 1, endonuclease (Drosophila)
Gene Aliases:
5830483C08Rik
Chromosome Location:
Chr.12: 11239718 - 11266029 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00724026_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gen1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177331.4 NP_796305.3 6 - 7 934 99
GenBank mRNA AK030995.1 - 6 - 7 944 99
AK031077.1 - 6 - 7 913 99
AK158255.1 - 5 - 6 757 99
BC090653.1 - 6 - 7 934 30659763 99
BC099936.1 - 5 - 6 743 99
BC138157.1 - 6 - 7 821 8861176 99
BC138158.1 - 6 - 7 821 8861177 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gen1

Panther Classification:

Molecular Function -

exodeoxyribonuclease

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
regulation of centrosome duplication
resolution of recombination intermediates
resolution of mitotic recombination intermediates
positive regulation of mitotic cell cycle spindle assembly checkpoint
nucleic acid phosphodiester bond hydrolysis
DNA binding
catalytic activity
nuclease activity
endonuclease activity
crossover junction endodeoxyribonuclease activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0