This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00723296_m1
See other IWS1 GE Assays ›
Gene Symbol:
Iws1
Gene Name:
IWS1 homolog (S. cerevisiae)
Gene Aliases:
1700069O15Rik, 3000008H23, AL117927, AW214353
Chromosome Location:
Chr.18: 32067325 - 32104331 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00723296_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Iws1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173441.3 NP_775617.1 7 - 8 1828 108
GenBank mRNA AK006925.1 - 1 - 2 121 108
AK028247.1 - 7 - 8 1799 108
AK033013.1 - 7 - 8 1823 108
AK136492.1 - 3 - 4 284 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000253.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Iws1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
transport
RNA splicing
regulation of mRNA export from nucleus
regulation of mRNA processing
mRNA transport
regulation of histone H4 acetylation
regulation of histone H3-K36 trimethylation
molecular_function
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800001800.mm.1 NM_173441.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001800.mm.1 NM_173441.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800001800.mm.1 TC1800000253.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm