This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712951_m1
See other ANTXR1 GE Assays ›
Gene Symbol:
Antxr1
Gene Name:
anthrax toxin receptor 1
Gene Aliases:
2310008J16Rik, 2810405N18Rik, Antrx1, Tem8
Chromosome Location:
Chr.6: 87133853 - 87335809 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712951_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Antxr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_054041.2 NP_473382.1 16 - 17 1683 82
GenBank mRNA AF378762.1 - 16 - 17 1623 82
AK013005.1 - 14 - 15 1090 82
AK031465.1 - 16 - 17 1673 82
BC057043.1 - 15 - 16 1147 82
BC094544.1 - 16 - 17 1628 6406827 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002648.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Antxr1

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
reproductive process
actin cytoskeleton reorganization
substrate adhesion-dependent cell spreading
receptor activity
transmembrane signaling receptor activity
collagen binding
metal ion binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600020592.mm.1 NM_054041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020592.mm.1 NM_054041.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600020592.mm.1 TC0600002648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0