This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712800_m1
See other RGS7BP GE Assays ›
Gene Symbol:
Rgs7bp
Gene Name:
regulator of G-protein signalling 7 binding protein
Gene Aliases:
A930030I01Rik, AI842293, D13Bwg1146e, R7bp
Chromosome Location:
Chr.13: 104945904 - 105054930 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712800_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs7bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029879.2 NP_084155.2 3 - 4 1179 108
GenBank mRNA AK020910.1 - 3 - 4 776 108
AK028323.1 - 3 - 4 556 108
AK032138.1 - 3 - 4 1179 108
AK036240.1 - 3 - 4 1086 108
AK046733.1 - 3 - 4 808 108
BC118957.2 - 3 - 4 711 40093423 108
DQ104214.1 - 3 - 4 469 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002606.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs7bp

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017754.mm.1 NM_029879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017754.mm.1 NM_029879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017754.mm.1 TC1300002606.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0