This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00685646_m1
See other ADGRF5 GE Assays ›
Gene Symbol:
Adgrf5
Gene Name:
adhesion G protein-coupled receptor F5
Gene Aliases:
8430401C09Rik, 9330185D23, AI528491, Gpr116, mKIAA0758
Chromosome Location:
Chr.17: 43360329 - 43459557 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00685646_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081178.1 NP_001074647.1 6 - 7 869 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

energy reserve metabolic process
regulation of lipid metabolic process
glucose homeostasis
surfactant homeostasis
fat cell differentiation
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0