Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100023694.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023694.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023694.mm.1 |
TC0100002950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023695.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023695.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023695.mm.1 |
TC0100002950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023692.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023692.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023692.mm.1 |
TC0100002950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023696.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023696.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023696.mm.1 |
TC0100002950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023697.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023697.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023697.mm.1 |
TC0100002950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023693.mm.1 |
NM_145506.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023693.mm.1 |
NM_145506.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023693.mm.1 |
TC0100002950.mm.1 |
MTA TC |