This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00660656_m1
See other SRRM4 GE Assays ›
Gene Symbol:
Srrm4
Gene Name:
serine/arginine repetitive matrix 4
Gene Aliases:
1500001A10Rik, B230202K19Rik, bv, mKIAA1853, nSR100
Chromosome Location:
Chr.5: 116439272 - 116592562 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00660656_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srrm4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026886.3 NP_081162.1 11 - 12 1738 87
GenBank mRNA AK053836.1 - 11 - 12 1470 87
AK129457.1 - 3 - 4 820 87
BC056392.1 - 11 - 12 1728 6855144 87
JN860221.1 - 11 - 12 1385 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003117.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srrm4

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
sensory perception of sound
RNA splicing
cell differentiation
regulation of RNA splicing
RNA binding
mRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500027753.mm.1 NM_026886.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027753.mm.1 NM_026886.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500027753.mm.1 TC0500003117.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0