This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00659801_m1
See other EIF3B GE Assays ›
Gene Symbol:
Eif3b
Gene Name:
eukaryotic translation initiation factor 3, subunit B
Gene Aliases:
AL033316, AL033334, AL033369, AW208965, D5Wsu45e, EIF3-ETA, EIF3-P110, EIF3-P116, Eif3s9, PRT1
Chromosome Location:
Chr.5: 140419305 - 140443358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00659801_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133916.2 NP_598677.1 13 - 14 1917 134
GenBank mRNA AK033722.1 - 13 - 14 1917 134
AK089978.1 - 13 - 14 1906 134
AK167618.1 - 13 - 14 1904 134
AK170938.1 - 13 - 14 1917 134
BC007175.1 - 12 - 13 1376 134
BC023767.1 - 13 - 14 1888 5343631 134
BC031704.1 - 13 - 14 1894 4023277 134
BC051065.1 - 13 - 14 1877 5343631 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001692.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3b

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translational initiation
nucleotide binding
nucleic acid binding
RNA binding
translation initiation factor activity
protein binding
translation initiation factor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500015557.mm.1 NM_133916.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015557.mm.1 NM_133916.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500015557.mm.1 TC0500001692.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015558.mm.1 NM_133916.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015558.mm.1 NM_133916.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500015558.mm.1 TC0500001692.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 926171 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0