This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00653140_g1
See other SORD GE Assays ›
Gene Symbol:
Sord
Gene Name:
sorbitol dehydrogenase
Gene Aliases:
Sdh-1, Sdh1, Sodh-1
Chromosome Location:
Chr.2: 122234839 - 122265337 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00653140_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sord
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146126.4 NP_666238.1 2 - 3 148 67
GenBank mRNA AK004692.1 - 2 - 3 172 67
AK015059.1 - 2 - 3 149 67
AK152169.1 - 2 - 3 197 67
AK166988.1 - 2 - 3 148 67
AK166996.1 - 2 - 3 177 67
AK209251.1 - 1 - 2 34 67
BC024124.1 - 2 - 3 163 5099395 67
BC030875.1 - 2 - 3 137 4237164 67
BC039957.1 - 2 - 3 148 67
BC092291.1 - 2 - 3 151 5097505 67
U27014.1 - 148 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sord

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

sorbitol metabolic process
sorbitol catabolic process
response to hormone
sperm motility
fructose biosynthetic process
response to cadmium ion
response to copper ion
L-xylitol catabolic process
L-xylitol metabolic process
oxidation-reduction process
L-iditol 2-dehydrogenase activity
zinc ion binding
oxidoreductase activity
identical protein binding
metal ion binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0