This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00628542_m1
See other TBCE GE Assays ›
Gene Symbol:
Tbce
Gene Name:
tubulin-specific chaperone E
Gene Aliases:
2610206D02Rik, C530005D02Rik, pmn
Chromosome Location:
Chr.13: 13997949 - 14039638 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00628542_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tbce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK011899.1 - 1 - 2 82 136
AK082940.1 - 1 - 2 102 136
AK160447.1 - 1 - 2 81 136
AK167383.1 - 1 - 2 90 136
AK167848.1 - 1 - 2 86 136
AY082332.1 - 1 - 2 56 136
BC050206.1 - 1 - 2 64 30009463 136

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK167383.1, AY082332.1, BC050206.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300009386.mm.1 NM_178337.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300009386.mm.1 TC1300001471.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300009385.mm.1 NM_178337.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300009385.mm.1 TC1300001471.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0