This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00628414_g1
See other SLC38A2 GE Assays ›
Gene Symbol:
Slc38a2
Gene Name:
solute carrier family 38, member 2
Gene Aliases:
5033402L14Rik, AI316867, ATA2, SAT2, mKIAA1382
Chromosome Location:
Chr.15: 96687386 - 96699724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00628414_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc38a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175121.3 NP_780330.2 13 - 14 1544 58
GenBank mRNA AK031615.1 - 13 - 14 1575 58
AK033812.1 - 1 - 2 84 58
AK129342.1 - 13 - 14 1505 58
AK164568.1 - 7 - 8 5307 58
AK169378.1 - 3 - 4 303 58
BC037675.1 - 10 - 11 951 58
BC041108.1 - 13 - 14 1414 4038546 58
BC048178.1 - 13 - 14 1544 6825801 58
BC049271.1 - 6 - 7 567 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002142.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc38a2

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
ion transport
sodium ion transport
amino acid transport
glycine betaine transport
alanine transport
amino acid transmembrane transporter activity
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500017189.mm.1 NM_175121.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500017189.mm.1 NM_175121.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500017189.mm.1 TC1500002142.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0