This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00628279_m1
See other ZFP280D GE Assays ›
Gene Symbol:
Zfp280d
Gene Name:
zinc finger protein 280D
Gene Aliases:
A930005F02Rik, BC027163, Suhw4, ZNF634, Znf280d
Chromosome Location:
Chr.9: 72274893 - 72363779 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00628279_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp280d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146224.4 NP_666336.3 16 - 17 2206 105
GenBank mRNA AK045173.1 - 16 - 17 2102 105
AK146597.1 - 16 - 17 2102 105
AK169622.1 - 15 - 16 2059 105
AK172652.1 - 18 - 19 2436 105
BC042445.1 - 17 - 18 2412 105
BC079878.1 - 16 - 17 2118 6855096 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000959.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp280d

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007386.mm.1 NM_146224.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900007386.mm.1 TC0900000959.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007382.mm.1 NM_146224.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900007382.mm.1 TC0900000959.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007383.mm.1 NM_146224.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900007383.mm.1 TC0900000959.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0