This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627682_m1
See other ATE1 GE Assays ›
Gene Symbol:
Ate1
Gene Name:
arginyltransferase 1
Gene Aliases:
AI225793, AW547406
Chromosome Location:
Chr.7: 130391493 - 130520371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627682_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ate1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029895.3 NP_001025066.1 6 - 7 1196 96
NM_013799.3 NP_038827.2 6 - 7 928 96
GenBank mRNA AK077471.1 - 2 - 3 347 96
AK148442.1 - 6 - 7 896 96
AK167591.1 - 6 - 7 888 96
BC042601.1 - 6 - 7 887 4503326 96
DQ093639.1 - 6 - 7 788 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004384.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ate1

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

protein arginylation
arginyltransferase activity
protein binding
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700036929.mm.1 NM_001029895.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036929.mm.1 NM_013799.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036929.mm.1 TC0700004384.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0