Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100039479.mm.1 |
NM_001112699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039479.mm.1 |
NM_001112699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039479.mm.1 |
NM_011180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039479.mm.1 |
NM_011180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039479.mm.1 |
TC1100004152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039476.mm.1 |
NM_001112699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039476.mm.1 |
NM_001112699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039476.mm.1 |
NM_011180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039476.mm.1 |
NM_011180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039476.mm.1 |
TC1100004152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039478.mm.1 |
NM_001112699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039478.mm.1 |
NM_001112699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039478.mm.1 |
NM_011180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039478.mm.1 |
NM_011180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039478.mm.1 |
TC1100004152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039477.mm.1 |
NM_001112699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039477.mm.1 |
NM_001112699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039477.mm.1 |
NM_011180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039477.mm.1 |
NM_011180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039477.mm.1 |
TC1100004152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_001112699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_001112699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_001112700.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_001112700.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_011180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
NM_011180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039471.mm.1 |
TC1100004152.mm.1 |
MTA TC |