This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627033_m1
See other GJB6 GE Assays ›
Gene Symbol:
Gjb6
Gene Name:
gap junction protein, beta 6
Gene Aliases:
AA958971, Cx30, D14Bwg0506e
Chromosome Location:
Chr.14: 57123301 - 57133611 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627033_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001010937.2 NP_001010937.1 1 - 2 429 111
NM_001271663.1 NP_001258592.1 1 - 2 429 111
GenBank mRNA AK134617.1 - 1 - 2 428 111
AK141610.1 - 1 - 2 429 111
BC013811.1 - 1 - 2 96 3483477 111
BC016507.1 - 1 - 2 104 3481872 111

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK134617.1, AK141610.1, BC013811.1, BC016507.1, NM_001010937.2, NM_001271663.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gjb6

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

cell communication
sensory perception of sound
negative regulation of cell proliferation
ear morphogenesis
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016476.mm.1 NM_001010937.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400016476.mm.1 NM_001271663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016476.mm.1 TC1400002141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016479.mm.1 NM_001010937.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400016479.mm.1 NM_001271663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016479.mm.1 TC1400002141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016474.mm.1 NM_001010937.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400016474.mm.1 NM_001271663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016474.mm.1 TC1400002141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016480.mm.1 NM_001010937.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400016480.mm.1 NM_001271663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016480.mm.1 TC1400002141.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 657996 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0